Learn More
Medchemexpress LLC Valencene | 4630-07-3 | 1 ML

Supplier: Medchemexpress LLC HYN663610MM/1ML
Valencene is a sesquiterpene isolated from Cyperus rotundus known for its antiallergic, antimelanogenesis, anti-inflammatory, and antioxidant activities. It functions by blocking the NF-κB pathway, inhibiting the expression of Th2 and proinflammatory chemokines, as well as the production of IL-1β and IL-6 in LPS-stimulated RAW 264.7 cells.
- Exhibits antiallergic, antimelanogenesis, anti-inflammatory, and antioxidant activities
- Inhibits expression of Th2 and proinflammatory chemokines
- Blocks the NF-κB pathway
- Reduces atopic dermatitis symptoms
- Restores decreased filaggrin expression in DNCB-sensitized mouse models
- Purity: 81.81%
- Molecular weight: 204.35
- Formula: C15H24
- Appearance: Colorless to light yellow liquid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.