Learn More
Cayman Chemical ValerylLcarnItInchLRIde 10mg
SDP
Supplier:  Cayman Chemical 2656310 MG
Encompass_Preferred
A short-chain acylcarnitine; levels increase in the serum of rhesus monkeys following exposure to 7 and 10 Gy of ionizing radiation
Catalog No. 50-293-3846