Learn More
Apexbio Technology LLC Valganciclovir HCl 175865-59-5 100mg
SDP
Supplier:  Apexbio Technology LLC B1864100
Encompass_Preferred
Valganciclovir hydrochloride is an antiviral prodrug that undergoes rapid enzymatic conversion to the active metabolite ganciclovir after oral administration Ganciclovir inhibits viral replication by selectively targeting viral DNA polymerase thereby interfering with viral DNA synthesis and slowing cytomegalovirus (CMV) proliferation Valganciclovir is commonly utilized in laboratory studies to evaluate antiviral activity against CMV and related herpesviruses and to investigate its therapeutic potential in CMV-associated infections including retinitis and organ transplant-associated CMV infections In vitro studies report an IC50 value for valganciclovir-derived ganciclovir ranging approximately from 0 2 to 2 8 M depending on experimental cell type and viral strain
Catalog No. 50-000-62016