Learn More
TARGETMOL CHEMICALS INC Valganciclovir hydrochloride
SDP
Supplier:  TARGETMOL CHEMICALS INC T1533100MG
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Valganciclovir hydrochloride (Valganciclovir HCl) is a hydrochloride salt form of valganciclovir, a prodrug form of ganciclovir, a nucleoside analog of 2'-deoxyguanosine, with antiviral activity. After phosphorylation, valganciclovir is incorporated into DNA, resulting in inhibition of viral DNA polymerase, and viral replication. Purity 98.55%
Catalog No. 50-287-9718