Learn More
Analytical Sales & Services Validator Chromatography Data System Validation Instrument
Supplier:  Analytical Sales & Services VALIDATOR
Encompass
High Resolution Signal/Peak Generator for Use with Chromatography Data Systems and Integrators. Includes External Power Supply and Documentation.
Catalog No. 50-008-06654