Learn More
Cayman Chemical Valinomycin Fatty Acids and De
SDP
Supplier:  Cayman Chemical 1000915210 MG
Encompass_Preferred
A cyclododecadepsipeptide potassium-selective ionophore antibotic; induces apoptosis in several cell tyes, including CHO cells, by stimulating potassium efflux
Catalog No. 50-289-1302