Learn More
Crescent Chemical Co Inc VALINOMYCIN RESEARCH GRADE
SDP
Supplier:  Crescent Chemical Co Inc SE38076.02
Encompass_Preferred
Valinomycin research grade Cyclopeptide-antibiotic. Affects potassium permeability of biomembranes. D-HIV = Hydroxy-isovaleric acid, Lac = lactic acid. Size - 10MG Storage Conditions - +15 °C TO +30 °C Catalog Number - 38076.02 CAS 200
Catalog No. 50-248-282