Learn More
Medchemexpress LLC Valiolamine | 83465-22-9 | 98.0% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY131114
Encompass_Preferred
Valiolamine is an aminocyclitol and an alpha-glucosidase inhibitor. It exhibits potent alpha-glucosidase inhibitory activity against porcine intestinal sucrase, maltase, and isomaltase. In vitro, it inhibits α-Glucosidase (Yeast), Maltase (Porcine), isomaltase (Porcine), sucrose (Porcine), and β-Glucosidase (Almond) with specific IC50 values.
  • Potent alpha-glucosidase inhibitor
  • Active against porcine intestinal sucrase, maltase, and isomaltase
  • Binds to porcine intestinal maltase and sucrase
  • Inhibits various alpha- and beta-glucosidases
Catalog No. 50-003-42076