Learn More
Beckman Coulter Valita Titer IgG Quantification Assay, 100-2000 mg/L (96-well) - Beckman Coulter VAL004
Supplier: Beckman Coulter VAL004
The Valita Titer assay is a rapid, plate-based IgG quantification assay that measures IgG concentrations in the range of 100-2000 mg/L using fluorescence polarization for a fast add-and-read workflow compatible with standard FP-equipped plate readers.
- Detection range: 100-2000 mg/L.
- Format: 96-well plate.
- Assay type: fluorescence polarization.
- Intended use: quantitative measurement of IgG in samples.
- Workflow: rapid add-and-read, suitable for high-throughput screening.
- Compatibility: works with plate readers that have a fluorescence polarization module.
Description
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.