Learn More
Beckman Coulter Valita Titer IgG Quantification Assay, 96-Well (2.5-100 mg/L) - Beckman Coulter VAL003
Supplier:  Beckman Coulter VAL003
Encompass
The Valita Titer assay is a rapid, plate-based method for quantifying IgG in high-throughput screening and clone selection workflows. It delivers fast, add-and-read results using fluorescence polarization detection and is optimized for low-range IgG measurement.
  • Rapid, add-and-read workflow for high-throughput screening.
  • Fluorescence polarization detection for IgG quantification.
  • Measurement range 2.5-100 mg/L.
  • 96-well plate format for plate-based assays.
  • Compatible with plate readers that have a fluorescence polarization module.
  • Suitable for clone screening and antibody titer measurement.
Catalog No. NC2331629
Only null left
Add to Cart
Request a Quote