Learn More
Medchemexpress LLC Valnemulin hydrochloride | 133868-46-9 | 98.1% | C31H53ClN2O5S | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0027100MG
Encompass_Preferred
Valnemulin hydrochloride is an orally effective truncated pleurotin antibiotic that inhibits protein synthesis in bacteria by binding to peptidyl transferase in the 50s ribosome subunit.
  • Effective in eliminating Mycobacterium bovis from the lungs in experimental bovine models.
  • Reduces mortality associated with epidemic rabbit enteropathy without adversely affecting growth performance.
  • Controls experimental infection in calves, diminishing clinical signs, restoring appetite, and reversing weight loss.
  • Reduces the isolation of Pasteurella multocida from infected calves' lungs.
Catalog No. 50-004-33427