Learn More
Medchemexpress LLC Valnivudine hydrochloride | 956483-03-7 | 98.99% | 534.04 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1985610MM/1ML
Encompass_Preferred
FV-100 is a potent, selective, and orally active anti-varicella zoster virus agent. It is a proagent of CF-1743 and demonstrates very low toxicity *in vivo*.
  • Potent anti-varicella zoster virus agent
  • Selective activity
  • Orally active
  • Proagent of CF-1743
  • Very low toxicity *in vivo*
Catalog No. 50-004-03309