Learn More
Cayman Chemical ValnoctamIde 10mg
SDP
Supplier:  Cayman Chemical 1845610 MG
Encompass_Preferred
An isomer of valpromide that has been marketed as an anxiolytic and sedative compound; abolishes the activity of MIP synthase in human brain crude homogenates (Ki = 0.18 mM); suppresses electrographic seizures in animal models of status epilepticus
Catalog No. 50-296-5986