Learn More
Medchemexpress LLC Valomaciclovir stearate | 195156-77-5 | 97.0% | 618.85 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY192881MG
Encompass_Preferred
Valomaciclovir stearate (ABT 606) is a nucleoside analog and the prodrug of Omaciclovir. It exhibits antiviral activity against HSV-1 and varicella zoster virus (VZV).
  • Nucleoside analog
  • Omaciclovir prodrug
  • Antiviral activity against HSV-1
  • Antiviral activity against varicella zoster virus (VZV)
Catalog No. 50-003-85854