Learn More
Medchemexpress LLC Valopicitabine dihydrochloride | 640725-71-9 | MFCD34589916 | 98.0% | 429.30 g/mol | C15H26Cl2N4O6 | 1 ML

Supplier: Medchemexpress LLC HY108060A10MM 1ML
Valopicitabine dihydrochloride is the dihydrochloride salt of valopicitabine (NM-283), a 3-O-valine ester prodrug of the nucleoside analog 2′-C-methylcytidine with activity against hepatitis C virus (HCV). The material is supplied as a white to off-white solid and is available as pre-made research stock solutions (for example, 10 mM in DMSO, 1 mL). Product documentation provides a stock solution preparation table and guidance for preparing a range of concentrations for research use.
- White to off-white solid form.
- Purity 98.0%.
- Molecular weight 429.30 g/mol.
- CAS number 640725-71-9.
- Available as 10 mM solution in DMSO (1 mL) for convenience.
- Soluble in DMSO and water.
- Includes stock solution preparation table for multiple concentrations.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.