Learn More
Medchemexpress LLC HY-10585 5g Medchemexpress, Valproic acid VPA;2-Propylpentanoic Acid CAS:99-66-1 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY105855G
Encompass_Preferred
Medchemexpress,HY-10585 5g Valproic acid VPA;2-Propylpentanoic Acid CAS:99-66-1 Formula:C8H16O2 Valproic acid is an HDAC inhibitor, with IC50 in the range of 0.5 and 2 mM, also inhibits HDAC1 (IC50, 400 μM), and induces proteasomal degradation of HDAC2; Valproic acid sodium salt is used in the treatment of epilepsy, bipolar disorder and prevention of migraine headaches. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-7296