Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Valproic acid-d15 | 362049-65-8 | MFCD01861597 | 99.8% | 159.30 | C8HD15O2 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY10585S25MG

Encompass_Preferred

Valproic acid-d15 is a deuterium-labeled stable isotope of valproic acid used as an internal standard and tracer for analytical, pharmacokinetic, and metabolic studies. It is supplied as a high-purity liquid suitable for LC-MS/MS and other mass spectrometry applications.

  • High isotopic enrichment with 99.8% purity.
  • Chemical formula C8HD15O2; molecular weight 159.30.
  • Available in small quantities for analytical workflows (5 MG).
  • Suitable for use as an internal standard in quantitative assays.
  • Storage recommendation: -20°C for long-term stability.

Catalog No. 50-002-43626


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.