Learn More
Medchemexpress LLC HY-10585A 500mg Medchemexpress, Valproic acid (sodium salt) CAS:1069-66-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10585A500MG
Encompass_Preferred
Medchemexpress, HY-10585A 500mg Valproic acid (sodium salt) CAS:1069-66-5 Valproic acid sodium salt (Sodium Valproate) is an HDAC inhibitor, with IC50 in the range of 0.5 and 2 mM, also inhibits HDAC1 (IC50, 400 μM), and induces proteasomal degradation of HDAC2. Valproic acid sodium salt activates Notch1 signaling and inhibits proliferation in small cell lung cancer (SCLC) cells. Valproic acid sodium salt is used in the treatment of epilepsy, bipolar disorder and prevention of migraine headaches. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8299