Learn More
Medchemexpress LLC Valsartan-d3 | 1331908-02-1 | 94.4% | 438.54 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY18204S11MG
Encompass_Preferred
Valsartan-d3 is the deuterium labeled Valsartan. Valsartan is an angiotensin II receptor antagonist and has the potential for high blood pressure and heart failure research. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Angiotensin II receptor antagonist
  • Potential for high blood pressure and heart failure research
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration may affect pharmacokinetic and metabolic profiles
Catalog No. 50-003-87618