Learn More
MSC Value Collection Dial Drop Indicator, 1.000 in Max Measurement, 0.0005 in Graduation, 0-50 Dial Reading
Supplier:  MSC 56450547
Encompass
This dial drop indicator provides precise measurements essential for industrial applications. Featuring a 1.000 inch maximum range and a 0.0005 inch graduation, it ensures accuracy for critical dimensional checks. The indicator includes a clear white dial with a 0-50 reading, complemented by a lug back for stable mounting.
  • Maximum measurement of 1.000 inch
  • Fine dial graduation of 0.0005 inch
  • Clear 0-50 dial reading
  • White dial for enhanced visibility
  • Integrated revolution counter
  • Lug back design for secure attachment
  • Utilizes English unit of measure
Catalog No. 50-005-31841