Learn More
MSC Value Collection Gage Block, Rectangle, Steel, 0.6 in, Grade AS-1
Supplier:  MSC 86294907
Encompass
This rectangular gage block is made of steel, measuring 0.6 inches, and features an accuracy grade of AS-1. It is suitable for precision measurement applications and comes with NIST certification.
  • Made of steel
  • Features accuracy grade AS-1
  • Measures 0.6 in
  • Rectangular shape
  • Includes NIST certification
Catalog No. 50-005-30387