Learn More
MSC Value Collection Gage Block Set, Steel, Grade 0, 81 Pc, 0.05 to 4 in Range
Supplier:  MSC 09904558
Encompass
This gage block set provides precise measurements for calibration and inspection tasks. Constructed from durable steel, the set features 81 pieces with an accuracy grade of 0, suitable for a wide range of applications from 0.05 to 4 inches.
  • Rectangle shape
  • Constructed from steel
  • Accuracy grade 0
  • Includes 81 individual pieces
  • Measures from 0.0500 to 4.0000 inch
  • Each block is individually serialized
  • Comes with a fitted case for storage
  • Meets ASME B89.1.9-2002 standards
Catalog No. 50-005-29231