Learn More
MSC Value Collection Gage Block, Square, 0.5 in, Grade 0, Steel
Supplier: MSC 86274701
This gage block is designed for precise measurements and calibration. It features a square shape and is constructed from steel, ensuring durability and stability. With an accuracy grade of 0, this block provides reliable precision, and each unit includes NIST certification for guaranteed quality and traceability in various applications.
- Square shape for versatile use
- Accuracy grade 0 for high precision
- Constructed from durable steel
- 0.5 in size for specific measurement requirements
- Includes NIST certification for quality assurance