Learn More
MSC Value Collection High Test Chain, 3/8 Inch, 5400 Lb Capacity, Zinc Plated
Supplier:  MSC 66344060
Encompass
This Welded High Test Chain from Value Collection is a robust and reliable solution for various demanding applications. It is engineered for durability and strength, featuring a high capacity and a corrosion-resistant finish, making it suitable for environments where heavy-duty performance and longevity are crucial. The chain is designed for specific length requirements as it is available in 1-foot increments from a 400-foot total coil length.
  • Constructed from grade 43 chain.
  • Features a durable zinc plated finish.
  • Offers a substantial 5,400 Lb capacity.
  • Available in 1-foot increments.
  • Total coil length of 400 feet.
  • Inside width is 0.5710 inches.
  • Inside length is 1.3590 inches.
Catalog No. 50-005-31194