Learn More
MSC Value Collection Steel Gage Block, Rectangle, 0.108 in, AS-1 Grade
Supplier:  MSC 86291457
Encompass
This product is a rectangular steel gage block designed for precise measurements. It comes with an AS-1 accuracy grade and is suitable for various industrial and laboratory applications requiring high precision.
  • Rectangular shape
  • AS-1 accuracy grade
  • Gage block type
  • Size of 0.108 in
  • Made from steel
  • Complies with NIST standards
  • Includes NIST certification
Catalog No. 50-005-30484