Learn More
Trajan Scientific VICI Valco Valco 6 Port Injection Valve, Large Bore Prep FLow Body (VAL.DLC6W-K-CTC)
Supplier:  Trajan Scientific 100.LC6PREP
Encompass
VICI Valco Valve 6 Port 2 Position, Prep FLow Body, 5K PSI, 0.75mm Ports. [VICI Valco PN: DLC6W-K-CTC] (NEW LPP PN: VAL.DLC6W-K-CTC). Made in the USA.
Catalog No. 50-635-062