Learn More
AVANTOR MASTERFLEX (VWR LLC) Parker Hannifin PP6VUC6-MG Ball Valve, Polypropylene, Union Connector 3/8'' push-to-connect tube, 1/each
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0137967
Encompass
This ball valve is glass reinforced polypropylene body with a polysulfone ball and EPDM seals. It is NSF 51 listed. It also is FDA compliant for contact with food. The plastic design is leak free and corrosion resistant.
Catalog No. NC0397284