Learn More
Medchemexpress LLC HY-109017 10mg , Vamorolone CAS:13209-41-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10901710MG
Encompass_Preferred
HY-109017 10mg Vamorolone CAS: 13209-41-1 Vamorolone (VBP15) is a first-in-class, orally active dissociative steroidal anti-inflammatory drug and membrane-stabilizer. Vamorolone improves muscular dystrophy without side effects. Vamorolone shows potent NF-κB inhibition and substantially reduces hormonal effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0497