Learn More
MSC Van Keuren Thread Measuring Wire, 72 TPI, 0.0139 in Pitch, 2 in Long
Supplier:  MSC 09186396
Encompass
This 3-piece set of thread measuring wires is designed for accurate measurement of thread pitch diameter. Each wire features 72 threads per inch (TPI) and a 0.0139 inch pitch, with a length of 2 inches.
  • Designed for thread pitch diameter measurement
  • 72 threads per inch (TPI)
  • 0.0139 inch pitch
  • 2 inch wire length
  • Nominal best wire diameter of 0.008 inch
  • Nominal constant of 0.012 inch
  • Supplied as a 3-piece set
Catalog No. 50-005-29568