Learn More
Strem, An Ascensus Company CAS# 12036-21-4. 50g. Vanadium (IV) oxide, 97% (99%-V). MFCD00049702
SDP
Supplier:  Strem, An Ascensus Company 93230950G
Encompass
CAS# 12036-21-4. 50g. Vanadium (IV) oxide, 97% (99%-V). MFCD00049702. Molecular Weight: 82.94. Molecular Formula: VO2. Color/form: black pwdr. Strem# 93-2309. http://www.strem.com/catalog/v/93-2309/
Catalog No. 50-901-14507