Learn More
Strem, An Ascensus Company CAS# 1314-62-1. 25g. Vanadium(V) oxide (99.99%-V) PURATREM. MFCD00011457
SDP
Supplier:  Strem, An Ascensus Company 93233725G
Encompass
CAS# 1314-62-1. 25g. Vanadium(V) oxide (99.99%-V) PURATREM. MFCD00011457. Molecular Weight: 181.88. Molecular Formula: V2O5. Color/form: -22 mesh orange pwdr. Strem# 93-2337. http://www.strem.com/catalog/v/93-2337/
Catalog No. 50-901-14522