Learn More
Perkin Elmer US LLC Vanadium (V) Pure Standard, 10,000 µg/mL, 15% HNO3, 125 mL
Supplier:  Perkin Elmer US LLC N9304356
Encompass_Preferred
Each Single Element Calibration Standard for AA and ICP-OES is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.
Catalog No. 50-211-1795