Learn More
Strem, An Ascensus Company CAS# 13709-31-4. 25g. Vanadium(V) trifluoride oxide, 98+%. MFCD00042555
SDP
Supplier:  Strem, An Ascensus Company 93233225G
Encompass
CAS# 13709-31-4. 25g. Vanadium(V) trifluoride oxide, 98+%. MFCD00042555. Molecular Weight: 123.94. Molecular Formula: VOF3. Color/form: yellow to orange pwdr. Strem# 93-2332. http://www.strem.com/catalog/v/93-2332/
Catalog No. 50-901-14515