Learn More
Selleck Chemical LLC Vancomycin HCl 50mg 1404-93-9
SDP
Supplier:  Selleck Chemical LLC S257550MG
Encompass_Preferred
Vancomycin HCl is a hydrochloride of vancomycin that is a narrow-spectrum glycopeptide antibacterial agent. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1177382