Learn More
Gold Biotechnology Inc Vancomycin Hydrochloride, USP Grade 1 g
SDP
Supplier:  Gold Biotechnology Inc V2001
Encompass_Preferred
Vancomycin hydrochloride is a member of the glycopeptide antibiotic class derived from Streptomyces orientalis. It is effective against gram-positive bacteria and used in studies of antibiotic resistance bacteria and nanoparticle transport studies. Exhibiting low toxicity to plants, vancomycin is suitable for plant selection and prevention of bacterial contamination in plant cell culture applications.
Catalog No. 50-153-2846