Learn More
Medchemexpress LLC Vancomycin (Standard) | 1404-90-6 | 95.9% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0671R5MG
Encompass_Preferred
Vancomycin (Standard) is the analytical standard of Vancomycin, intended for research and analytical applications. It is an antibiotic used for the treatment of bacterial infections. The compound is an analytical standard and is commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for Vancomycin
  • Intended for research and analytical applications
  • Antibiotic for bacterial infections
  • Used in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS)
Catalog No. 50-004-07709