Learn More
Medchemexpress LLC Neboglamine hydrochl 50mg | 2759182-59-5 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY114753A50MG
Encompass_Preferred
Neboglamine hydrochloride is an orally active positive allosteric modulator of the NMDA receptor glycine site, used in schizophrenia and cognitive research. Supplied for laboratory research use only as a solid or as a DMSO solution.
  • Orally active NMDA receptor glycine-site positive modulator.
  • Used in schizophrenia and cognitive function research.
  • Available as solid and 10 mM solution formats in DMSO.
  • Typical pack sizes include 5 MG, 10 MG, 25 MG, 50 MG, and 100 MG.
  • High purity (~97.0%) with white to off-white solid appearance.
  • Storage: powder at -20°C (long term) or 4°C (short term); in solvent at -80°C or -20°C.
Catalog No. 50-002-29295