Learn More
Medchemexpress LLC Vanillyl alcohol | 498-00-0 | 154.16 | 10 G
SDP
Supplier:  Medchemexpress LLC HYN206710G
Encompass_Preferred
Vanillyl alcohol is an orally active phenolic alcohol that reduces ROS generation, suppresses Bax, and increases Bcl-2. It exhibits anti-angiogenic, anti-inflammatory, anti-nociceptive, and neuroprotective effects, and is also used as a flavoring agent in foods and beverages.
  • Reduces ROS generation
  • Suppresses Bax and increases Bcl-2
  • Exhibits anti-angiogenic, anti-inflammatory, anti-nociceptive, and neuroprotective effects
  • Used as a flavoring agent in foods and beverages
  • Increases viability of MPP+-induced MN9D dopaminergic cells
  • Inhibits acetic acid-induced vascular permeability and writhing response in mice
  • Reduces exudate and inflammation in carrageenan-induced air pouches in mice
Catalog No. 50-004-14960