Learn More
Medchemexpress LLC Vanillyl alcohol | 498-00-0 | 154.16 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYN2067
Encompass_Preferred
Vanillyl alcohol (p-(Hydroxymethyl)guaiacol) is an orally active phenolic alcohol that reduces ROS generation, suppresses Bax, and increases Bcl-2. It exhibits anti-angiogenic, anti-inflammatory, anti-nociceptive, and neuroprotective effects. Vanillyl alcohol is also utilized as a flavoring agent in foods and beverages.
  • Reduces ROS generation.
  • Suppresses Bax.
  • Increases Bcl-2.
  • Exhibits anti-angiogenic effects.
  • Exhibits anti-inflammatory effects.
  • Exhibits anti-nociceptive effects.
  • Exhibits neuroprotective effects.
  • Utilized as a flavoring agent in foods and beverages.
Catalog No. 50-003-89518