Learn More
Medchemexpress LLC Vanillyl alcohol | 498-00-0 | 154.16 | 50 G
SDP
Supplier:  Medchemexpress LLC HYN206750G
Encompass_Preferred
Vanillyl alcohol, also known as p-(Hydroxymethyl)guaiacol, is an orally active phenolic alcohol. It is recognized for its ability to decrease reactive oxygen species (ROS) generation and modulate apoptotic pathways by suppressing Bax and increasing Bcl-2. This compound also functions as a flavoring agent in various foods and beverages.
  • Reduces ROS generation.
  • Suppresses Bax.
  • Increases Bcl-2.
  • Possesses anti-angiogenic effects.
  • Exhibits anti-inflammatory properties.
  • Offers anti-nociceptive effects.
  • Provides neuroprotective benefits.
  • Used as a flavoring agent in foods and beverages.
Catalog No. 50-004-15231