Learn More
Medchemexpress LLC Vanillyl butyl ether | 82654-98-6 | MFCD00238529 | 99.8% | 210.27 g/mol | C12H18O3 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW014394
Encompass_Preferred
Vanillyl butyl ether is a research-grade small molecule used as a repellent agent and a probe in studies of transient receptor potential (TRP) channels. It is provided at high purity with documented solubility and storage data to support in vitro and in vivo formulation and long-term stability.
Catalog No. 50-003-04114