Learn More
Medchemexpress LLC Vanillyl butyl ether | 82654-98-6 | MFCD00238529 | 99.8% | 210.27 g/mol | C12H18O3 | 50 G

Supplier: Medchemexpress LLC HYW01439450G
Vanillyl butyl ether is a vanillyl derivative provided as a high-purity research chemical used as a mild warming agent and repellent. It is used in formulation development, sensory testing, and biological assays exploring warming or repellent activity.
- High purity (99.75%), suitable for research applications.
- Chemical formula C12H18O3 and molecular weight 210.27 g/mol.
- Soluble in DMSO and common formulation vehicles at ≥2.5 mg/mL (≈11.9 mM).
- Stable when stored under recommended conditions (pure form: -20°C/4°C; in solvent: -80°C/-20°C).
- Available in multiple pack sizes for assay and formulation needs.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.