Learn More
Medchemexpress LLC Vanillylmandelic acid | 55-10-7 | 99.8% | 198.17 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY113121S
Encompass_Preferred
Vanillylmandelic acid is the end-product of epinephrine and norepinephrine metabolism. It can be used as an indication of disorder in neurotransmitter metabolism and exhibits antioxidant activity towards the DPPH radical.
  • Indicates disorders in neurotransmitter metabolism
  • Exhibits antioxidant activity against the DPPH radical
Catalog No. 50-004-39407