Learn More
Medchemexpress LLC Vanilpyruvic acid | 1081-71-6 | MFCD00056707 | 98.3% | 210.18 g·mol^-1 | C10H10O5 | 1 ML

Supplier: Medchemexpress LLC HY10141610MM 1ML
Vanilpyruvic acid is a catecholamine metabolite and precursor to vanillactic acid. It is supplied for analytical and biochemical research applications, available both as ready-to-use solutions and as solid reference material for assay development and metabolic studies.
- High purity: 98.3%.
- Molecular formula C10H10O5; molecular weight 210.18 g·mol^-1.
- CAS number 1081-71-6.
- Available as 10 mM solution in DMSO (1 ML) and as solid sizes from 5 MG to 500 MG.
- Suitable as an analytical standard and for studies of catecholamine metabolism.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.