Learn More
Medchemexpress LLC Vanin-1-IN-3 | 2940964-70-3 | 96.2% | 377.36 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14641250MG
Encompass_Preferred
Vanin-1-IN-3 (OMP-7) is a vanin-1 inhibitor with an IC50 of 0.038 μM. For research use only, not for sale to patients.
  • IC50: 0.038 μM (vanin-1)
  • Exhibits potent activity against both serum and renal vanin-1 in vivo (hamsters) at 10 mg/kg.
Catalog No. 50-003-60747