Learn More
Medchemexpress LLC Vanin-1-in-3 | 2940964-70-3 | 96.2% | C17H22F3NO5 | 10MG

Supplier: Medchemexpress LLC HY14641210MG
Vanin-1-IN-3 (OMP-7) is a small-molecule inhibitor of vanin-1 for biochemical and preclinical research. It shows potent activity in vitro and has documented formulation protocols for in vivo studies, with reported high purity and solubility guidance for experimental use.
- Potent vanin-1 inhibition (IC50 0.038 μM).
- Suitable for in vitro biochemical assays and cellular studies.
- Formulation examples provided for in vivo dosing protocols.
- High reported purity (96.2%).
- Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
- Appearance: colorless to light yellow oil.
- Storage stability documented for long-term and in-solvent conditions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.