Learn More
Electron Microscopy Sciences MicroPoint™ Featherlite Vannas Scissors Curved
SDP
Supplier:  Electron Microscopy Sciences 77900C
Encompass_Preferred
MicroPoint Featherlite  Style DC-1 has curved Vannas Scissors has 5mm smooth blades.
OAL: 125 mm
Catalog No. 50-306-26