Learn More
Cayman Chemical VardenafIl-d5 500ug
SDP
Supplier:  Cayman Chemical 25225500 UG
Encompass_Preferred
An internal standard for the quantification of vardenafil by GC- or LC-MS
Catalog No. 50-293-6995