Learn More
Medchemexpress LLC Vardenafil-d5 hydrochloride | 224785-91-5 | 98.7% | 530.09 g/mol | C23H28D5ClN6O4S | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0442AS10MG
Encompass_Preferred
Deuterium-labeled vardenafil hydrochloride supplied as a powder for use as an analytical reference and research standard. It is the hydrochloride salt of vardenafil with deuterium labeling (d5), provided in small milligram quantities for analytical and method development work.
  • Deuterated internal standard for analytical assays.
  • High purity suitable for reference use.
  • Molecular weight 530.09 g/mol, consistent with d5 hydrochloride.
  • Supplied as a powder in small package sizes for laboratory use.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-97997