Learn More
TARGETMOL CHEMICALS INC VARDENAFIL HYDROCHLORIDE 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T409725MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Vardenafil hydrochloride trihydrate (BAY38-9456) is a new type PDE inhibitor with IC50 of 0.7 and 180 nM for PDE5 and PDE1 respectively. purity: 99%
Catalog No. 50-257-6810